Learn More
Supply Solutions Blue Levo Manual Pipette Filler 1 - 100mL
Catalog No. NC3166540
Encompass_Preferred
Levo is an easy to use serological pipette controller for use with 1-100mL serological pipettes. Simply attach a serological pipette into the silicone holder and squeeze the bulb (which creates a vacuum) and you are ready to pipette. The thumb lever controls both the aspirate and dispense speed for up to 50mL per squeeze. A press of the central button blows-out any remaining drops in the pipette. An easily replaceable 3µm hydrophobic filter prevents liquids from contaminating the inside of the Levo.Comfortable designPrecise pipetting controlAutoclavable silicone pipette holder
Catalog No. NC3166540 Supplier Supply Solutions Supplier No. 740100019999
Levo is an easy to use serological pipette controller for use with 1-100mL serological pipettes. Simply attach a serological pipette into the silicone holder and squeeze the bulb (which creates a vacuum) and you are ready to pipette. The thumb lever controls both the aspirate and dispense speed for up to 50mL per squeeze. A press of the central button blows-out any remaining drops in the pipette. An easily replaceable 3µm hydrophobic filter prevents liquids from contaminating the inside of the Levo.Comfortable designPrecise pipetting controlAutoclavable silicone pipette holder