Learn More
Lime Media Manual Pump Plastic Dispenser
Catalog No. 19043983
Change view
Click to view available options
Quantity:
15/Cs.
Catalog No. Quantity
19-043-983 15/Cs.
1 options
Catalog No. 19-043-983 Supplier Lime Media Supplier No. 197775WPM
Choose a manual pump hand sanitizing dispenser made of high-grade white ABS plastic, bulk refillable 34 oz. tank, fluid level viewing window, and non-drip pump mechanism.
  • White ABS plastic construction
  • 34 oz. bulk-fill tank
  • Wall mount applicable
  • All hardware included
  • For use of soap or gel sanitizer
  • Sanitizing fluid viewing window
Specifications
Product Type Manual Pump Sanitizing Dispenser
Quantity 15/Cs.
Sterility Non-sterile