Learn More
Grainger Mitutoyo Manual-Reading Protractor, Stainless Steel, 6 in., 1° Graduation, 1 ea.
Catalog No. 502793575
Encompass_Preferred
This manual-reading protractor features a rectangular head and is designed for precise angle measurement in woodworking and metalworking applications. The device utilizes a 180° arc to allow for quick and accurate alignment between two workpieces.
  • Features a 6 in. stainless steel blade with 1 degree graduations.
  • Includes a satin chrome finish on the scale plate and indicator blade for enhanced readability.
  • Designed with a rectangular square head.
  • Capable of measuring angles from 0° to 180°.
  • Equipped with a magnetic head for secure positioning.
  • Mechanical operation type.
Catalog No. 50-279-3575 Supplier Grainger Supplier No. 1ARY7
This manual-reading protractor features a rectangular head and is designed for precise angle measurement in woodworking and metalworking applications. The device utilizes a 180° arc to allow for quick and accurate alignment between two workpieces.
  • Features a 6 in. stainless steel blade with 1 degree graduations.
  • Includes a satin chrome finish on the scale plate and indicator blade for enhanced readability.
  • Designed with a rectangular square head.
  • Capable of measuring angles from 0° to 180°.
  • Equipped with a magnetic head for secure positioning.
  • Mechanical operation type.