Learn More
MSC Vestil Mobile Hand Lift Table, 220 lb Capacity, 1 ea
Catalog No. NC1811947
Encompass
This hydraulic elevating cart is designed for transporting heavy loads in laboratory, food service, and healthcare environments. It is not suitable for wash-down applications. Key components like the platform, handle, base frame, caster rig, and hardware are 304-grade stainless steel for durability and corrosion resistance. The pump assembly, scissor legs, and foot pedal have a silver painted finish for maximum rust-resistance. The platform is raised with a foot-operated hydraulic pump and lowered with a hand-operated release, equipped with poly-on-poly wheels. Each cylinder includes an internal safety velocity fuse for enhanced safety.
  • Unique lift design quickly raises and lowers material
  • Ergonomic cart design improves user safety
  • Caster construction provides easy portability when loaded
  • Partially stainless steel construction offers long-lasting rugged use
Catalog No. NC1811947 Supplier MSC Supplier No. 91444745
This hydraulic elevating cart is designed for transporting heavy loads in laboratory, food service, and healthcare environments. It is not suitable for wash-down applications. Key components like the platform, handle, base frame, caster rig, and hardware are 304-grade stainless steel for durability and corrosion resistance. The pump assembly, scissor legs, and foot pedal have a silver painted finish for maximum rust-resistance. The platform is raised with a foot-operated hydraulic pump and lowered with a hand-operated release, equipped with poly-on-poly wheels. Each cylinder includes an internal safety velocity fuse for enhanced safety.
  • Unique lift design quickly raises and lowers material
  • Ergonomic cart design improves user safety
  • Caster construction provides easy portability when loaded
  • Partially stainless steel construction offers long-lasting rugged use