Learn More
Analytical Sales & Services Manual SS 2 Position, 6 Port Injector with Switch
Catalog No. 5000807028
Encompass
Stainless Steel 2 Position, 6 Port Injector with Switch - 600 bar\n\nQty/Pkg: EACH
Catalog No. 50-008-07028 Supplier Analytical Sales & Services Supplier No. 47725I1884
Stainless Steel 2 Position, 6 Port Injector with Switch - 600 bar\n\nQty/Pkg: EACH