Learn More
Analytical Sales & Services Manual SS 2 Position, 6 Port Sample Injector with Attached 20µL Sample Loop
Catalog No. 5000807030
Encompass
Manual Stainless Steel 2 Position, 6 Port Standard Analytical Syringe Load Sample Injector with Attached 20µL Sample Loop\nInject from 1µL to 5mL, Adjustable to 7,000psi\n\nQty/Pkg: EACH
Catalog No. 50-008-07030 Supplier Analytical Sales & Services Supplier No. 477254
Manual Stainless Steel 2 Position, 6 Port Standard Analytical Syringe Load Sample Injector with Attached 20µL Sample Loop\nInject from 1µL to 5mL, Adjustable to 7,000psi\n\nQty/Pkg: EACH