Learn More
World Precision Instrument Manual Stereotaxic Injector
SDP
Catalog No. 502539157
Encompass
For precisely-controlled infusion/withdrawal of picoliter volumes
Stereotaxic Injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control.
Mounted directly on a stereotaxic frame, it allows precise injection directly through the needle into the tissue. This avoids the compliance issues that arise when injections are forced through plastic tubing. Volumes as small as 200 picoliters may be infused.
Catalog No. 50-253-9157 Supplier World Precision Instrument Supplier No. 505258
For precisely-controlled infusion/withdrawal of picoliter volumes
Stereotaxic Injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control.
Mounted directly on a stereotaxic frame, it allows precise injection directly through the needle into the tissue. This avoids the compliance issues that arise when injections are forced through plastic tubing. Volumes as small as 200 picoliters may be infused.