Learn More
Grainger Vestil Manual Trolley, Steel, 1,000 Lb. Capacity, 1 Ea.
Catalog No. 502777221
Encompass_Preferred
These adjustable steel beam trolleys provide a reliable solution for suspending and positioning hoists on S, H, W, and M type I-beams. Designed for quick installation and manual adjustment, they are ideal for use in maintenance shops, warehouses, fabrication areas, manufacturing plants, loading zones, and material handling workstations requiring controlled horizontal load movement along an overhead beam.
  • Capacity of 1,000 lb.
  • Compatible with beam flange widths ranging from 2 in. to 4-1/2 in.
  • Manual screw mechanism with locking ring for fast and simple width adjustments
  • Constructed with four sealed bearing rollers for smooth operation
  • Includes an integrated lifting eye for secure hoist attachment
Catalog No. 50-277-7221 Supplier Grainger Supplier No. 172Y44
These adjustable steel beam trolleys provide a reliable solution for suspending and positioning hoists on S, H, W, and M type I-beams. Designed for quick installation and manual adjustment, they are ideal for use in maintenance shops, warehouses, fabrication areas, manufacturing plants, loading zones, and material handling workstations requiring controlled horizontal load movement along an overhead beam.
  • Capacity of 1,000 lb.
  • Compatible with beam flange widths ranging from 2 in. to 4-1/2 in.
  • Manual screw mechanism with locking ring for fast and simple width adjustments
  • Constructed with four sealed bearing rollers for smooth operation
  • Includes an integrated lifting eye for secure hoist attachment