Learn More
Grainger Pneumadyne, Inc. Manual Air Control Valve, Three-Way, Two-Position, Button, 1 ea
Catalog No. 502753542
Encompass_Preferred
This manual air control valve is a three-way, two-position, normally closed device designed for precision airflow management. Featuring a durable push-button actuator with a spring return, this component is vacuum operable and suitable for panel mounting applications.
  • 3-way, 2-position, normally closed switching function
  • 1/8 in and 10-32 female NPT port connections
  • Maximum operating pressure of 125 psi
  • Temperature range of -20°F to 160°F
  • 0.24 coefficient of volume (Cv)
  • Corrosion-resistant construction featuring aluminum, brass, and Buna-N seals
  • Non-threaded exhaust port
Catalog No. 50-275-3542 Supplier Grainger Supplier No. 14F266
This manual air control valve is a three-way, two-position, normally closed device designed for precision airflow management. Featuring a durable push-button actuator with a spring return, this component is vacuum operable and suitable for panel mounting applications.
  • 3-way, 2-position, normally closed switching function
  • 1/8 in and 10-32 female NPT port connections
  • Maximum operating pressure of 125 psi
  • Temperature range of -20°F to 160°F
  • 0.24 coefficient of volume (Cv)
  • Corrosion-resistant construction featuring aluminum, brass, and Buna-N seals
  • Non-threaded exhaust port