Learn More
BURKLE INC Manually-operated reel EX with grounding cable
Catalog No. 5000025969
Encompass_Preferred
The manually-operated drum reels can be equipped with various cables or chains.
If flammable or highly flammable liquids are to be collected, then electrically conductive manually-operated drum reels must be used together with a grounding cable. The dipping bottles must be made of non-sparking metal.
Catalog No. 50-000-25969 Supplier BURKLE INC Supplier No. 53090030
The manually-operated drum reels can be equipped with various cables or chains.
If flammable or highly flammable liquids are to be collected, then electrically conductive manually-operated drum reels must be used together with a grounding cable. The dipping bottles must be made of non-sparking metal.