Learn More
Cayman Chemical Manumycin A, 52665-74-4, 1 mg
SDP
Supplier:  Cayman Chemical 100104971
Encompass_Preferred
A potent and selective farnesyltransferase (FTase) inhibitor with anti-tumor activity: inhibits rat brain FTase with a Ki value of 1.2 µM, thereby preventing Ras activation; inhibits IKK in an number of cells types
Catalog No. NC9890454