Learn More
Cayman Chemical Manumycin A Inhibitors
Supplier: Cayman Chemical 100104975 MG
A potent and selective farnesyltransferase (FTase) inhibitor with anti-tumor activity: inhibits rat brain FTase with a Ki value of 1.2 µM, thereby preventing Ras activation; inhibits IKK in an number of cells types