Learn More
Cayman Chemical Manumycin A Inhibitors
SDP
Catalog No. 502888414
Encompass_Preferred
A potent and selective farnesyltransferase (FTase) inhibitor with anti-tumor activity: inhibits rat brain FTase with a Ki value of 1.2 µM, thereby preventing Ras activation; inhibits IKK in an number of cells types
Catalog No. 50-288-8414 Supplier Cayman Chemical Supplier No. 100104971 MG
A potent and selective farnesyltransferase (FTase) inhibitor with anti-tumor activity: inhibits rat brain FTase with a Ki value of 1.2 µM, thereby preventing Ras activation; inhibits IKK in an number of cells types