Learn More
Hach Company ManVer 2 Hardness Indicator Powder Pillows, pk/100
Supplier:  Hach Company 92899
Encompass
ManVer 2 (Calmagite) Hardness Indicator, for determination of hardness and chelants by titration. Pack of 100 powder pillows
Catalog No. 50-199-4470