Learn More
MSC Miller Manyard II Shock Absorbing Lanyard, Kevlar, 6 ft., 1 ea.
Catalog No. 5000509862
Encompass
This single-leg, shock-absorbing lanyard is designed to protect workers from falls in environments involving potential electric arc-flash and arc-blast exposure. Engineered for use at heights or in confined spaces near energized electrical sources, it features a specialized core that expands to reduce fall arrest forces. The lanyard is constructed from Kevlar® webbing and includes a choke-off loop with a locking snap hook.
  • Contracts from 6 ft. to 4 ft. to minimize tripping, snagging, and dragging
  • Features a unique warning flag that indicates if the device has been involved in a fall
  • Includes an integral bright red core for reliable visual inspection
  • Meets all applicable OSHA, ANSI, CSA, and ASTM F887-05 arc-flash standards
  • Provides a weight capacity of 310 lb.
  • Equipped with a 3/4 in. snap hook throat opening
Catalog No. 50-005-09862 Supplier MSC Supplier No. 59122598
This single-leg, shock-absorbing lanyard is designed to protect workers from falls in environments involving potential electric arc-flash and arc-blast exposure. Engineered for use at heights or in confined spaces near energized electrical sources, it features a specialized core that expands to reduce fall arrest forces. The lanyard is constructed from Kevlar® webbing and includes a choke-off loop with a locking snap hook.
  • Contracts from 6 ft. to 4 ft. to minimize tripping, snagging, and dragging
  • Features a unique warning flag that indicates if the device has been involved in a fall
  • Includes an integral bright red core for reliable visual inspection
  • Meets all applicable OSHA, ANSI, CSA, and ASTM F887-05 arc-flash standards
  • Provides a weight capacity of 310 lb.
  • Equipped with a 3/4 in. snap hook throat opening