Learn More
Enzo Life Sciences Manzamine A (1mg). CAS: 104196-68-1
SDP
Supplier:  Enzo Life Sciences ALX350294M001
Encompass_Preferred
β-Carboline alkaloid with anti-inflammatory, antimalarial, antifungal, anti-HIV-1 and insecticidal activity. Also active against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus. Inhibits GSK-3. Has native fluorescence, highest when solubilized in methanol (Ex: 340nm, Em: 387nm). Purity: ≥98% (HPLC). Appearance: Off white to yellow solid. Solubility: Soluble in DMSO, methanol or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8832