Learn More
Medchemexpress LLC MAO-B-IN-9 | 2416910-88-6 | 99.7% | 300.40 | 25 MG
SDP
Catalog No. 5000363479
Encompass_Preferred
MAO-B-IN-9 (compound 16) is a potent, selective, BBB-penetrated, irreversible, and time-dependent MAO-B (monoamine oxidase B) inhibitor with an IC50 of 0.18 μM. It prevents Aβ1-42-induced neuronal cell death and exhibits neuroprotective effects, potentially due to its Aβ1-42 anti-aggregation properties. MAO-B-IN-9 is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, selective, BBB-penetrated, irreversible, and time-dependent MAO-B inhibitor
  • Prevents Aβ1-42-induced neuronal cell death
  • Exhibits neuroprotective effects
  • Click chemistry reagent with alkyne group for CuAAc reactions
Catalog No. 50-003-63479 Supplier Medchemexpress LLC Supplier No. 25MGHY146314
MAO-B-IN-9 (compound 16) is a potent, selective, BBB-penetrated, irreversible, and time-dependent MAO-B (monoamine oxidase B) inhibitor with an IC50 of 0.18 μM. It prevents Aβ1-42-induced neuronal cell death and exhibits neuroprotective effects, potentially due to its Aβ1-42 anti-aggregation properties. MAO-B-IN-9 is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, selective, BBB-penetrated, irreversible, and time-dependent MAO-B inhibitor
  • Prevents Aβ1-42-induced neuronal cell death
  • Exhibits neuroprotective effects
  • Click chemistry reagent with alkyne group for CuAAc reactions