Learn More
Medchemexpress LLC HY-131036 5mg Medchemexpress, MAO-IN-M30 (dihydrochloride) CAS:64821-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310365MG
Encompass_Preferred
Medchemexpress, HY-131036 5mg MAO-IN-M30 (dihydrochloride) CAS:64821-19-8 MAO-IN-M30 dihydrochloride is an orally active, brain-permeable, and brain selective irreversible MAO-A (IC50=37 nM) and MAO-B (IC50=57 nM) inhibitor. MAO-IN-M30 dihydrochloride is a potent iron chelator and radical scavenger. MAO-IN-M30 dihydrochloride has a neuroprotective effect against Dexamethasone-induced brain cell apoptosis. MAO-IN-M30 dihydrochloride also exhibits neurorestorative activity in post MPTP and lactacystin models of Parkinson's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1258