Learn More
Medchemexpress LLC Mao-in-m30 dihydrochloride | 64821-19-8 | MFCD20488063 | 99.3% | C14H16Cl2N2O | 10MG
SDP
Catalog No. 5000261815
Encompass_Preferred
MAO-IN-M30 dihydrochloride is an orally active, brain-permeable, and brain-selective irreversible inhibitor of monoamine oxidase A and B (MAO-A and MAO-B). It also acts as a potent iron chelator and radical scavenger, showing neuroprotective and neurorestorative effects in preclinical models, and contains an alkyne moiety suitable for click-chemistry labeling.
  • Orally active and brain-permeable inhibitor of MAO-A and MAO-B.
  • Potent inhibition (MAO-A IC50 = 37 nM; MAO-B IC50 = 57 nM).
  • Functions as an iron chelator and radical scavenger.
  • Demonstrates neuroprotective and neurorestorative effects in preclinical models.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Supplied as a dihydrochloride salt with high purity (~99.3%).
Catalog No. 50-002-61815 Supplier Medchemexpress LLC Supplier No. HY13103610MG
MAO-IN-M30 dihydrochloride is an orally active, brain-permeable, and brain-selective irreversible inhibitor of monoamine oxidase A and B (MAO-A and MAO-B). It also acts as a potent iron chelator and radical scavenger, showing neuroprotective and neurorestorative effects in preclinical models, and contains an alkyne moiety suitable for click-chemistry labeling.
  • Orally active and brain-permeable inhibitor of MAO-A and MAO-B.
  • Potent inhibition (MAO-A IC50 = 37 nM; MAO-B IC50 = 57 nM).
  • Functions as an iron chelator and radical scavenger.
  • Demonstrates neuroprotective and neurorestorative effects in preclinical models.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Supplied as a dihydrochloride salt with high purity (~99.3%).