Learn More
Aviva Systems Biology Corp MAOA Antibody - middle region (ARP56075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAOA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAOA. Alternate names: MAO-AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61709