Learn More
Medchemexpress LLC Maohuoside A | 128988-55-6 | 1 MG
SDP
Catalog No. 5000420575
Encompass_Preferred
Maohuoside A is a single compound isolated from E. koreanum that potently promotes osteogenesis. It enhances the osteogenesis of bone marrow-derived mesenchymal stem cells via bone morphogenetic protein (BMP) and MAPK signaling pathways.
  • For research use only
  • Not sold to patients
Catalog No. 50-004-20575 Supplier Medchemexpress LLC Supplier No. HYN40191MG
Maohuoside A is a single compound isolated from E. koreanum that potently promotes osteogenesis. It enhances the osteogenesis of bone marrow-derived mesenchymal stem cells via bone morphogenetic protein (BMP) and MAPK signaling pathways.
  • For research use only
  • Not sold to patients