Learn More
Medchemexpress LLC Maohuoside A | 128988-55-6 | MFCD31697553 | 5 MG
SDP
Catalog No. 5000419249
Encompass_Preferred
Maohuoside A is a single compound isolated from *E. koreanum* that potently promotes osteogenesis. It enhances the osteogenesis of bone marrow-derived mesenchymal stem cells via bone morphogenetic protein (BMP) and MAPK signaling pathways.
  • Potently promotes osteogenesis
  • Enhances osteogenesis of bone marrow-derived mesenchymal stem cells
  • Works via bone morphogenetic protein (BMP) and MAPK signaling pathways
Catalog No. 50-004-19249 Supplier Medchemexpress LLC Supplier No. HYN40195MG
Maohuoside A is a single compound isolated from *E. koreanum* that potently promotes osteogenesis. It enhances the osteogenesis of bone marrow-derived mesenchymal stem cells via bone morphogenetic protein (BMP) and MAPK signaling pathways.
  • Potently promotes osteogenesis
  • Enhances osteogenesis of bone marrow-derived mesenchymal stem cells
  • Works via bone morphogenetic protein (BMP) and MAPK signaling pathways