Learn More
TARGETMOL CHEMICALS INC Maohuoside A 1MG
SDP
Catalog No. 502879658
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Maohuoside A promotes osteogenesis of rat mesenchymal stem cells via BMP and MAPK signaling pathways. Purity 98.95%
Catalog No. 50-287-9658 Supplier TARGETMOL CHEMICALS INC Supplier No. TN20811MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Maohuoside A promotes osteogenesis of rat mesenchymal stem cells via BMP and MAPK signaling pathways. Purity 98.95%