Learn More
Enzo Life Sciences [pThr183, pTyr185] MAP kinase (177-189) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1640001
Encompass_Preferred
Diphosphopeptide with sequence, DHTGFLpTEpYVATR, corresponding to MAP kinase residues 177-189 inclusive of putative activation sites. Peptide can be used as a VHR phosphatase substrate (BML-SE333). Phosphate release may be quantified using BML-AK111 malachite green colorimetric reagent. Phosphate release is much more rapid for the phosphotyrosine residue compared to the phosphothreonine residue, both in vitro and in vivo. Purity: ≥95% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-201-1269