Learn More
ECM Bioscience MAP Kinase Activation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068752
Encompass_Preferred
The MAP kinase activation antibody sampler kit can be used to examine the activation of three major MAPKs: p38, ERK, and JNK. The kit also includes antibodies to monitor total expression levels of p38, ERK1/2, and JNK1. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8752 Supplier ECM Bioscience Supplier No. MK6050
The MAP kinase activation antibody sampler kit can be used to examine the activation of three major MAPKs: p38, ERK, and JNK. The kit also includes antibodies to monitor total expression levels of p38, ERK1/2, and JNK1. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms