Learn More
Meso Scale Discovery MAP Kinase WCL20ate
SDP
Catalog No. 5000081783
Encompass
The MAP Kinase Whole Cell Lysate Kit from Meso Scale Discovery provides reagents to detect total and phosphorylated ERK1/2 JNK1/2 and p38 MAP kinases in cell lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies and buffers needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a wide dynamic range This kit allows for the quantitative analysis of MAP kinase activation in cells With a single 96-well plate up to 40 samples can be analyzed in triplicate
Catalog No. 50-000-81783 Supplier Meso Scale Discovery Supplier No. K15101D3
Add to Cart
Add to Cart
Request a Quote
The MAP Kinase Whole Cell Lysate Kit from Meso Scale Discovery provides reagents to detect total and phosphorylated ERK1/2 JNK1/2 and p38 MAP kinases in cell lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies and buffers needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a wide dynamic range This kit allows for the quantitative analysis of MAP kinase activation in cells With a single 96-well plate up to 40 samples can be analyzed in triplicate