Learn More
Meso Scale Discovery MAP Kinase Whole Cell Lysate
SDP
Catalog No. 50917444
Encompass
The MAP Kinase Whole Cell Lysate Kit from Meso Scale Discovery contains reagents to lyse cells and analyze MAP kinase (ERK/p38/JNK) phosphorylation in cell lysates The kit includes lysis buffer phosphatase inhibitors and blocking reagents optimized for electrochemiluminescent immunoassays This allows for efficient cell lysis and sample preparation to measure MAP kinase activation in cells The kit contains sufficient reagents to prepare lysates from approximately 500 microplates of adherent cells or 1000 tubes of suspension cells for use in Meso Scale Discovery assays
This item is not returnable. View return policy
Catalog No. 50-917-444 Supplier Meso Scale Discovery Supplier No. K15101D2
Add to Cart
Add to Cart
Request a Quote
The MAP Kinase Whole Cell Lysate Kit from Meso Scale Discovery contains reagents to lyse cells and analyze MAP kinase (ERK/p38/JNK) phosphorylation in cell lysates The kit includes lysis buffer phosphatase inhibitors and blocking reagents optimized for electrochemiluminescent immunoassays This allows for efficient cell lysis and sample preparation to measure MAP kinase activation in cells The kit contains sufficient reagents to prepare lysates from approximately 500 microplates of adherent cells or 1000 tubes of suspension cells for use in Meso Scale Discovery assays