Learn More
Sino Biological MAP1D Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10883T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MAP1D (rh MAP1D; Catalog#10883-H08B; NP_954697.1; Arg44-Ala335). MAP1D specific IgG was purified by Human MAP1D affinity chromatography.
Catalog No. 50-224-8579