Learn More
Aves Labs, Inc Anti-Microtubule-Associated Protein (MAP2) Antibody
Supplier:  Aves Labs, Inc MAP
Encompass
Concentration: 200 g/mL; Clonality: Polyclonal; Host Species: Chicken; Species Reactivity: Human, Mouse, Rat; Applications: ICC, IHC, WB; Protein Name / Synonyms: Microtubule-associated protein 2 (MAP-2)
Catalog No. NC1651838