Learn More
ABclonal Technology MAP2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2220520UL
Encompass_Preferred
This gene encodes a protein that belongs to the microtubule-associated protein family The proteins of this family are thought to be involved in microtubule assembly which is an essential step in neurogenesis The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dentrites implicating a role in determining and stabilizing dentritic shape during neuron development A number of alternatively spliced variants encoding distinct isoforms have been described
Catalog No. 50-000-05930