Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MAP2K1 (MEK1), His Tag Recombinant Protein

Catalog No. PR5043A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR5043A 100 μg
PV3303 10 μg
PV4003 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR5043A Supplier Invitrogen™ Supplier No. PR5043A
Only null left
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

MAP2K1 (MEK1) kinase is a dual-specificity protein kinase that functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation. MAP2K1 is activated by a wide variety of growth factors and cytokines and also by membrane-dependent depolarization and calcium influx. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02750
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.05% Triton X-100, 2mM DTT, 0.05mM EDTA, 150mM NaCl, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5604
Molecular Weight (g/mol) 49.2 kDa
Name Human MAP2K1 (MEK1), His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias CFC3; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; Map2k1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg
Common Name MEK1
Gene Symbol Map2k1
KinaseFamily STE7 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: STE Group
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Signal Transduction
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.