Learn More
Aviva Systems Biology Corp MAP2K2 Antibody - C-terminal region (ARP48942_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48942_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP2K2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MAP2K2. Alternate names: MAPKK2, MEK2, MKK2, PRKMK2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-59968