Learn More
Aviva Systems Biology Corp MAP2K3 Antibody - C-terminal region (ARP42065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP2K3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MAP2K3. Alternate names: MAPKK3, MEK3, MKK3, PRKMK3, SAPKK2, SAPKK-2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58096