Learn More
Aviva Systems Biology Corp Map2k5 Antibody - N-terminal region (ARP49021_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49021_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Map2k5. It was validated on Western Blot. . Alternate names: AI324775, AI428457, MEK5, Mapkk5, Prkmk5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-393