Learn More
Carnabio Usa Inc MAP2K7 INACTIVE /50UG/FULL-LEN
Supplier:  Carnabio Usa Inc 071471050
Encompass
MAP2K7[inactive], Full-length, Wild type, Amino Acid 1-419, NP_660186.1, Expressed in E. coli
Catalog No. 50-754-1045