Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4214MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 2F6. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: A431, HeLa or HL-60 cells or liver tissue. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Partial recombinant MAP3K1 (aa1211-1310) (SKNSMTLDLNSSSKCDDSFGCSSNSSNAVIPSDETVFTP-VEEKCRLDVNTELNSSIEDLLEASMPSSDTTVTFKSEVAVLSPEKAENDDTYKDDVNHNQK). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 195kDa (intact); 80kDa (cleaved).Chromosome: 5q11.2. Gene Symbol: MAP3K1. Gene ID: 4214. Ensemble: ENSG00000095015. Uniprot: XP_016864973.
Catalog No. 50-188-4928