Learn More
Aviva Systems Biology Corp MAP3K1 Antibody - C-terminal region (ARP48684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP3K1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MAP3K1. Alternate names: MAPKKK1, MEKK, MEKK1, SRXY6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59885