Learn More
Aviva Systems Biology Corp MAP3K10 Antibody - C-terminal region (ARP60309_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60309_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP3K10. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MAP3K10. Alternate names: MEKK10, MLK2, MSTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-001