Learn More
Aviva Systems Biology Corp MAP3K11 Antibody - middle region (ARP56349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP3K11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAP3K11. Alternate names: MGC17114, MLK-3, MLK3, PTK1, SPRK, MEKK11ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61824