Learn More
Carnabio Usa Inc MAP3K6/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 071085
Encompass
MAP3K6, Catalytic domain, Wild type, Amino Acid 622-950, NP_004663.3, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0503