Learn More
Aviva Systems Biology Corp MAP3K8 Antibody - C-terminal region (ARP30649_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30649_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP3K8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MAP3K8. Alternate names: COT, EST, ESTF, TPL2, MEKK8, Tpl-2, c-COTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52822