Learn More
Novus Biologicals™ MAP3K8/Tpl2/COT Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 1326 |
| Species | Human |
| Purity | >85%, by HPLC |
| Concentration | 2.5 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-62178 |
| Gene Alias | Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 |
| Gene Symbol | MAP3K8 |
| Quantity | 0.25 mg |
| Research Category | Angiogenesis, Cancer, Protein Kinase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.