Learn More
Aviva Systems Biology Corp MAP4 Antibody - N-terminal region (ARP60314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MAP4. Alternate names: DKFZp779A1753, MGC8617ApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-287-002