Learn More
Medchemexpress LLC MAP4343 25mg | 511-26-2 | 25MG
SDP
Catalog No. 5000239859
Encompass_Preferred
MAP4343 is the 3-methylether derivative of Pregnenolone MAP4343 binds in vitro to microtubule-associated protein 2 (MAP2) stimulates the polymerization of tubulin enhances the extension of neurites and protects neurons against neurotoxic agents[1 [2
Catalog No. 50-002-39859 Supplier Medchemexpress LLC Supplier No. HY10711625MG
MAP4343 is the 3-methylether derivative of Pregnenolone MAP4343 binds in vitro to microtubule-associated protein 2 (MAP2) stimulates the polymerization of tubulin enhances the extension of neurites and protects neurons against neurotoxic agents[1 [2