Learn More
Carnabio Usa Inc MAP4K2/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0711110
Encompass
MAP4K2, Full-length, Wild type, Amino Acid 1-820 , NP_004570.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0863