Learn More
Aviva Systems Biology Corp MAP4K4 Antibody - N-terminal region (ARP49031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP4K4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAP4K4. Alternate names: FLH21957, FLJ10410, FLJ20373, FLJ90111, HGK, KIAA0687, NIK, MEKKK4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59998