Learn More
Aviva Systems Biology Corp MAP4K5 Antibody - middle region (ARP33101_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP4K5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAP4K5. Alternate names: GCKR, KHS, KHS1, MAPKKKK5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54885