Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ AdvanTech™ StanSolv™ A-10CR Nitrile Gloves

Catalog No. 113927E Shop All MAPA Professional Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-392-7E 11
11-392-7A 7
11-392-7B 8
11-392-7C 9
11-392-7D 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11-392-7E Supplier MAPA™ Supplier No. 519311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Z-pattern grip nitrile gloves are used for semiconductor wet processes using solvent-based strippers, etchants, and cleaners. Also used for handling cleanroom and clean process acids, caustics and solvents in ISO 4/Class 100 environment.

Ideal for polysilicone handling, pure chemicals manufacturing, solvents handling and chemical mixing and handling.

  • Outperform natural rubber gloves with organic solvents and caustics commonly used in electronics
  • Offer excellent resistance to cuts, punctures, and abrasions
  • Unique “Z” pattern embossed grip for secure handling of wet or dry objects
  • Hand specific
  • Thickness: 11mil
  • Length: 13 in. (33cm)
  • Green

Specifications

Cleanroom Class Class 10 (ISO 4)
Color Green
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nitrile
Size 11
Thickness (English) 11 mil
Length (English) 13 in.
Length (Metric) 33 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.