Description
- Butyl protection for chemical gases and liquids, such as ketones, esters and amines
- Ideal for use with paints, resins, varnish and inks
- Optimal embossed textured grip
- Comfortable, unlined medium weight glove
- Rolled cuff to catch drips and facilitate attachment to chemical suits
Specifications
Specifications
| Certifications/Compliance | CE EUR CAT 3, EN 388 0111 |
| Certifications/Compliance (Europe) | EN 16350, Cat 3 CE 0334 |
| Color | Black |
| Latex Free | Yes |
| Material | Butyl |
| Size | 7 |
| Thickness (English) | 20 mil |
| Thickness (Metric) | 0.50 mm |
| EN388 Standard | EN 388:0010X |
| EN ISO 374-1 | Type A |
| Show More |