Learn More
MSC MAPA Professional Chemical Resistant Gloves, Jersette 303, Medium, 1 ea
Catalog No. 5000793985
Encompass
These Type B chemical-resistant gloves, model Jersette 303, are constructed from FSC® certified latex with a textile support lining. They are designed for enhanced dexterity and provide resistance against snags, abrasions, cuts, and punctures.
  • Constructed from FSC® certified latex with textile support
  • Blue color, 12 inch overall length
  • Pebbled grip surface for enhanced dexterity
  • Thickness of 45 mil (1.15 mm)
  • Supported for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Scalloped cuff style, seamless design
  • Certified to EN 407 X1XXXX; EN ISO 374-1/Type B KPT; EN 388 2131X; CAT III 0334 standards
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level 1
  • ANSI/ISEA puncture resistance level 1
Catalog No. 50-007-93985 Supplier MSC Supplier No. 87538344
These Type B chemical-resistant gloves, model Jersette 303, are constructed from FSC® certified latex with a textile support lining. They are designed for enhanced dexterity and provide resistance against snags, abrasions, cuts, and punctures.
  • Constructed from FSC® certified latex with textile support
  • Blue color, 12 inch overall length
  • Pebbled grip surface for enhanced dexterity
  • Thickness of 45 mil (1.15 mm)
  • Supported for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Scalloped cuff style, seamless design
  • Certified to EN 407 X1XXXX; EN ISO 374-1/Type B KPT; EN 388 2131X; CAT III 0334 standards
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level 1
  • ANSI/ISEA puncture resistance level 1